Assay Details
| Gene Symbol: LOC4335977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778429.2 | XP_015633915.1 | 3 - 4 | 1125 | 73 | |
| GenBank mRNA | AB056061.1 | - | 3 - 4 | 1085 | 73 | |
| AK068340.1 | - | 3 - 4 | 1111 | 73 | ||
More Information
Set Membership: |
|

LOC4335977
Genomic Map