Assay Details
| Gene Symbol: LOC4336362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780807.2 | XP_015636293.1 | 22 - 23 | 4462 | 113 | |
| XM_015780808.2 | XP_015636294.1 | 20 - 21 | 3854 | 113 | ||
| GenBank mRNA | AB353922.1 | - | 22 - 23 | 4467 | 113 | |
| AK066586.1 | - | 10 - 11 | 2644 | 113 | ||
| EU009924.1 | - | 23 - 24 | 4435 | 113 | ||
More Information
Set Membership: |
|

LOC4336362
Genomic Map