Assay Details
| Gene Symbol: LOC4336362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780807.2 | XP_015636293.1 | 13 - 14 | 1906 | 112 | |
| XM_015780808.2 | XP_015636294.1 | 11 - 12 | 1298 | 112 | ||
| GenBank mRNA | AB353922.1 | - | 13 - 14 | 1911 | 112 | |
| AK066586.1 | - | 1 - 2 | 88 | 112 | ||
| EU009924.1 | - | 14 - 15 | 1879 | 112 | ||
More Information
Set Membership: |
|

LOC4336362
Genomic Map