Assay Details
| Gene Symbol: LOC4336362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780807.2 | XP_015636293.1 | 18 - 19 | 3192 | 121 | |
| XM_015780808.2 | XP_015636294.1 | 16 - 17 | 2584 | 121 | ||
| GenBank mRNA | AB353922.1 | - | 18 - 19 | 3197 | 121 | |
| AK066586.1 | - | 6 - 7 | 1374 | 121 | ||
| EU009924.1 | - | 19 - 20 | 3165 | 121 | ||
More Information
Set Membership: |
|

LOC4336362
Genomic Map