Assay Details
| Gene Symbol: LOC4336362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780807.2 | XP_015636293.1 | 20 - 21 | 3869 | 87 | |
| XM_015780808.2 | XP_015636294.1 | 18 - 19 | 3261 | 87 | ||
| GenBank mRNA | AB353922.1 | - | 20 - 21 | 3874 | 87 | |
| AK066586.1 | - | 8 - 9 | 2051 | 87 | ||
| EU009924.1 | - | 21 - 22 | 3842 | 87 | ||
More Information
Set Membership: |
|

LOC4336362
Genomic Map