Assay Details
| Gene Symbol: LOC4336362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780807.2 | XP_015636293.1 | 21 - 22 | 4134 | 74 | |
| XM_015780808.2 | XP_015636294.1 | 19 - 20 | 3526 | 74 | ||
| GenBank mRNA | AB353922.1 | - | 21 - 22 | 4139 | 74 | |
| AK066586.1 | - | 9 - 10 | 2316 | 74 | ||
| EU009924.1 | - | 22 - 23 | 4107 | 74 | ||
More Information
Set Membership: |
|

LOC4336362
Genomic Map