Assay Details
| Gene Symbol: LOC4336391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781211.2 | XP_015636697.1 | 9 - 10 | 1401 | 60 | |
| GenBank mRNA | AK058333.1 | - | 2 - 3 | 371 | 60 | |
More Information
Set Membership: |
|

LOC4336391
Genomic Map