Assay Details
| Gene Symbol: LOC4336547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780081.2 | XP_015635567.1 | 7 - 8 | 1385 | 97 | |
| GenBank mRNA | AK068610.1 | - | 7 - 8 | 1345 | 97 | |
| AY224498.1 | - | 2 - 3 | 676 | 97 | ||
More Information
Set Membership: |
|

LOC4336547
Genomic Map