Assay Details
| Gene Symbol: LOC4337825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783895.1 | XP_015639381.1 | 21 - 22 | 3170 | 95 | |
| GenBank mRNA | AK100732.1 | - | 21 - 22 | 3177 | 95 | |
More Information
Set Membership: |
|

LOC4337825
Genomic Map