Assay Details
| Gene Symbol: LOC4339694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785335.2 | XP_015640821.1 | 1 - 2 | 771 | 71 | |
| GenBank mRNA | AK241368.1 | - | 1 - 2 | 464 | 71 | |
| AK287489.1 | - | 1 - 2 | 462 | 71 | ||
More Information
Set Membership: |
|

LOC4339694
Genomic Map