Assay Details
| Gene Symbol: LOC4340403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788594.1 | XP_015644080.1 | 1 | 1438 | 135 | |
| GenBank mRNA | AK106381.1 | - | 1 | 946 | 135 | |
More Information
Set Membership: |
|

LOC4340403
Genomic Map