Assay Details
| Gene Symbol: LOC4340503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788213.2 | XP_015643699.1 | 19 - 20 | 2217 | 70 | |
| XM_015788214.2 | XP_015643700.1 | 19 - 20 | 2217 | 70 | ||
| GenBank mRNA | AK065677.1 | - | 19 - 20 | 2216 | 70 | |
| AK100667.1 | - | 19 - 20 | 2216 | 70 | ||
More Information
Set Membership: |
|

LOC4340503
Genomic Map