Assay Details
| Gene Symbol: LOC4340751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788927.2 | XP_015644413.1 | 1 - 2 | 718 | 64 | |
| GenBank mRNA | AK066102.1 | - | 1 - 2 | 582 | 64 | |
| AK099920.1 | - | 1 - 2 | 646 | 64 | ||
| AK120497.1 | - | 1 - 2 | 714 | 64 | ||
More Information
Set Membership: |
|

LOC4340751
Genomic Map