Assay Details
| Gene Symbol: LOC4340981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787142.2 | XP_015642628.1 | 2 - 3 | 561 | 76 | |
| XM_015787143.2 | XP_015642629.1 | 2 - 3 | 561 | 76 | ||
More Information
Set Membership: |
|

LOC4340981
Genomic Map