Assay Details
| Gene Symbol: LOC4340981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787142.2 | XP_015642628.1 | 4 - 5 | 1057 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4340981| Gene Symbol: LOC4340981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787142.2 | XP_015642628.1 | 4 - 5 | 1057 | 66 | |
Set Membership: |
|