Assay Details
| Gene Symbol: LOC4342105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788607.1 | XP_015644093.1 | 3 - 4 | 436 | 74 | |
| GenBank mRNA | AK073747.1 | - | 3 - 4 | 445 | 74 | |
More Information
Set Membership: |
|

LOC4342105
Genomic Map