Assay Details
| Gene Symbol: LOC4343041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791884.2 | XP_015647370.1 | 3 - 4 | 709 | 93 | |
| GenBank mRNA | AK243062.1 | - | 3 - 4 | 574 | 93 | |
More Information
Set Membership: |
|

LOC4343041
Genomic Map