Assay Details
| Gene Symbol: LOC4343294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792302.1 | XP_015647788.1 | 10 - 11 | 1477 | 67 | |
| GenBank mRNA | AK061706.1 | - | 7 - 8 | 677 | 67 | |
| AK064866.1 | - | 10 - 11 | 1280 | 67 | ||
| AK065801.1 | - | 10 - 11 | 1376 | 67 | ||
More Information
Set Membership: |
|

LOC4343294
Genomic Map