Assay Details
| Gene Symbol: LOC4343294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792302.1 | XP_015647788.1 | 15 - 16 | 1878 | 81 | |
| GenBank mRNA | AK061706.1 | - | 12 - 13 | 1078 | 81 | |
| AK064866.1 | - | 15 - 16 | 1681 | 81 | ||
| AK065801.1 | - | 15 - 16 | 1777 | 81 | ||
More Information
Set Membership: |
|

LOC4343294
Genomic Map