Assay Details
| Gene Symbol: LOC4343294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792302.1 | XP_015647788.1 | 5 - 6 | 1012 | 80 | |
| GenBank mRNA | AK061706.1 | - | 2 - 3 | 212 | 80 | |
| AK064866.1 | - | 5 - 6 | 815 | 80 | ||
| AK065801.1 | - | 5 - 6 | 911 | 80 | ||
More Information
Set Membership: |
|

LOC4343294
Genomic Map