Assay Details
| Gene Symbol: LOC4343294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792302.1 | XP_015647788.1 | 7 - 8 | 1180 | 60 | |
| GenBank mRNA | AK061706.1 | - | 4 - 5 | 380 | 60 | |
| AK064866.1 | - | 7 - 8 | 983 | 60 | ||
| AK065801.1 | - | 7 - 8 | 1079 | 60 | ||
More Information
Set Membership: |
|

LOC4343294
Genomic Map