Assay Details
| Gene Symbol: LOC4343294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792302.1 | XP_015647788.1 | 8 - 9 | 1259 | 80 | |
| GenBank mRNA | AK061706.1 | - | 5 - 6 | 459 | 80 | |
| AK064866.1 | - | 8 - 9 | 1062 | 80 | ||
| AK065801.1 | - | 8 - 9 | 1158 | 80 | ||
More Information
Set Membership: |
|

LOC4343294
Genomic Map