Assay Details
| Gene Symbol: LOC4343294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792302.1 | XP_015647788.1 | 9 - 10 | 1366 | 78 | |
| GenBank mRNA | AK061706.1 | - | 6 - 7 | 566 | 78 | |
| AK064866.1 | - | 9 - 10 | 1169 | 78 | ||
| AK065801.1 | - | 9 - 10 | 1265 | 78 | ||
More Information
Set Membership: |
|

LOC4343294
Genomic Map