Assay Details
Gene Symbol: LOC4343299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791108.2 | XP_015646594.1 | 10 - 11 | 4871 | 116 | |
XM_015791109.2 | XP_015646595.1 | 10 - 11 | 5077 | 116 | ||
XM_015791110.2 | XP_015646596.1 | 10 - 11 | 4814 | 116 | ||
XM_015791111.2 | XP_015646597.1 | 10 - 11 | 4874 | 116 | ||
XM_015791112.2 | XP_015646598.1 | 10 - 11 | 4754 | 116 | ||
GenBank mRNA | AK100332.1 | - | 5 - 6 | 2586 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
