Assay Details
| Gene Symbol: LOC4324616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757368.2 | XP_015612854.1 | 18 - 19 | 2294 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4324616| Gene Symbol: LOC4324616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757368.2 | XP_015612854.1 | 18 - 19 | 2294 | 107 | |
Set Membership: |
|