Assay Details
| Gene Symbol: LOC4346383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755194.2 | XP_015610680.1 | 1 - 2 | 486 | 60 | |
| GenBank mRNA | AK063547.1 | - | 1 - 2 | 483 | 60 | |
More Information
Set Membership: |
|

LOC4346383
Genomic Map