Assay Details
| Gene Symbol: LOC4346383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755194.2 | XP_015610680.1 | 3 - 4 | 669 | 71 | |
| GenBank mRNA | AK063547.1 | - | 4 - 5 | 743 | 71 | |
More Information
Set Membership: |
|

LOC4346383
Genomic Map