Assay Details
| Gene Symbol: LOC4346383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755194.2 | XP_015610680.1 | 4 - 5 | 773 | 109 | |
| GenBank mRNA | AK063547.1 | - | 5 - 6 | 847 | 109 | |
More Information
Set Membership: |
|

LOC4346383
Genomic Map