Assay Details
| Gene Symbol: LOC4347161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795685.2 | XP_015651171.1 | 3 - 4 | 615 | 70 | |
| GenBank mRNA | AK061837.1 | - | 3 - 4 | 606 | 70 | |
| AK098903.1 | - | 3 - 4 | 587 | 70 | ||
| JX972160.1 | - | 3 - 4 | 577 | 70 | ||
More Information
Set Membership: |
|

LOC4347161
Genomic Map