Assay Details
| Gene Symbol: LOC4349659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761755.2 | XP_015617241.1 | 4 - 5 | 783 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4349659| Gene Symbol: LOC4349659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761755.2 | XP_015617241.1 | 4 - 5 | 783 | 80 | |
Set Membership: |
|