Assay Details
| Gene Symbol: LOC4349897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761409.1 | XP_015616895.1 | 1 - 2 | 287 | 56 | |
| GenBank mRNA | AK073758.1 | - | 1 - 2 | 238 | 56 | |
| AK104899.1 | - | 1 - 2 | 242 | 56 | ||
| JX972166.1 | - | 1 - 2 | 216 | 56 | ||
| KT862211.1 | - | 1 - 2 | 214 | 56 | ||
| Gene Symbol: LOC4345681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK105927.1 | - | 233 | 56 | ||
More Information
Set Membership: |
|

LOC4349897
Genomic Map