Assay Details
| Gene Symbol: LOC4349897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761409.1 | XP_015616895.1 | 2 - 3 | 554 | 110 | |
| GenBank mRNA | AK073758.1 | - | 2 - 3 | 505 | 110 | |
| AK104899.1 | - | 2 - 3 | 509 | 110 | ||
| JX972166.1 | - | 2 - 3 | 483 | 110 | ||
| KT862211.1 | - | 2 - 3 | 481 | 110 | ||
| Gene Symbol: LOC4345681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK105927.1 | - | 500 | 110 | ||
More Information
Set Membership: |
|

LOC4349897
Genomic Map