Assay Details
| Gene Symbol: LOC4349897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761409.1 | XP_015616895.1 | 3 - 4 | 667 | 92 | |
| GenBank mRNA | AK073758.1 | - | 3 - 4 | 618 | 92 | |
| AK104899.1 | - | 3 - 4 | 622 | 92 | ||
| JX972166.1 | - | 3 - 4 | 596 | 92 | ||
| KT862211.1 | - | 3 - 4 | 594 | 92 | ||
| Gene Symbol: LOC4345681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK105927.1 | - | 613 | 92 | ||
More Information
Set Membership: |
|

LOC4349897
Genomic Map