Assay Details
| Gene Symbol: LOC4351204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759619.2 | XP_015615105.1 | 1 | 171 | 85 | |
| GenBank mRNA | AK243460.1 | - | 1 | 85 | 85 | |
More Information
Set Membership: |
|

LOC4351204
Genomic Map