Assay Details
| Gene Symbol: LOC4323863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766032.2 | XP_015621518.1 | 16 - 17 | 2116 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4323863| Gene Symbol: LOC4323863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766032.2 | XP_015621518.1 | 16 - 17 | 2116 | 87 | |
Set Membership: |
|