Assay Details
| Gene Symbol: LOC4325162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758793.2 | XP_015614279.1 | 11 - 12 | 2687 | 106 | |
| XM_015758800.2 | XP_015614286.1 | 11 - 12 | 2684 | 106 | ||
| XM_015758806.2 | XP_015614292.1 | 11 - 12 | 2473 | 106 | ||
| XM_015758811.2 | XP_015614297.1 | 11 - 12 | 2578 | 106 | ||
| GenBank mRNA | AK068471.1 | - | 11 - 12 | 2479 | 106 | |
| AK072849.1 | - | 11 - 12 | 2275 | 106 | ||
More Information
Set Membership: |
|

LOC4325162
Genomic Map