Assay Details
| Gene Symbol: LOC4325322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786731.2 | XP_015642217.1 | 3 - 4 | 863 | 96 | |
| GenBank mRNA | AK243338.1 | - | 3 - 4 | 818 | 96 | |
More Information
Set Membership: |
|

LOC4325322
Genomic Map