Assay Details
| Gene Symbol: LOC4325106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794675.2 | XP_015650161.1 | 4 - 5 | 403 | 92 | |
| XM_015794687.2 | XP_015650173.1 | 4 - 5 | 402 | 92 | ||
| GenBank mRNA | AK243365.1 | - | 4 - 5 | 392 | 92 | |
More Information
Set Membership: |
|

LOC4325106
Genomic Map