Assay Details
| Gene Symbol: LOC4328116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770246.2 | XP_015625732.1 | 4 - 5 | 784 | 100 | |
| GenBank mRNA | AK069443.1 | - | 4 - 5 | 461 | 100 | |
| AK100842.1 | - | 4 - 5 | 790 | 100 | ||
More Information
Set Membership: |
|

LOC4328116
Genomic Map