Assay Details
| Gene Symbol: LOC4329057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767951.2 | XP_015623437.1 | 5 - 6 | 784 | 118 | |
| GenBank mRNA | AK101872.1 | - | 5 - 6 | 791 | 118 | |
More Information
Set Membership: |
|

LOC4329057
Genomic Map