Assay Details
| Gene Symbol: LOC4329449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770526.2 | XP_015626012.1 | 2 - 3 | 319 | 68 | |
| XM_015770527.2 | XP_015626013.1 | 2 - 3 | 319 | 68 | ||
| GenBank mRNA | AK058487.1 | - | 2 - 3 | 305 | 68 | |
| AK122107.1 | - | 2 - 3 | 246 | 68 | ||
More Information
Set Membership: |
|

LOC4329449
Genomic Map