Assay Details
| Gene Symbol: LOC4330499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770624.1 | XP_015626110.1 | 18 - 19 | 2553 | 93 | |
| XM_015770625.1 | XP_015626111.1 | 18 - 19 | 2552 | 93 | ||
| XM_015770627.1 | XP_015626113.1 | 17 - 18 | 2041 | 93 | ||
| XM_015770628.2 | XP_015626114.1 | 18 - 19 | 2516 | 93 | ||
| GenBank mRNA | AK065045.1 | - | 18 - 19 | 2518 | 93 | |
More Information
Set Membership: |
|

LOC4330499
Genomic Map