Assay Details
| Gene Symbol: LOC4330499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770624.1 | XP_015626110.1 | 23 - 24 | 3291 | 102 | |
| XM_015770625.1 | XP_015626111.1 | 22 - 23 | 3236 | 102 | ||
| XM_015770627.1 | XP_015626113.1 | 22 - 23 | 2779 | 102 | ||
| XM_015770628.2 | XP_015626114.1 | 22 - 23 | 3200 | 102 | ||
| GenBank mRNA | AK065045.1 | - | 22 - 23 | 3202 | 102 | |
More Information
Set Membership: |
|

LOC4330499
Genomic Map