Assay Details
| Gene Symbol: LOC4330499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770627.1 | XP_015626113.1 | 1 - 2 | 71 | 66 | |
| XM_015770628.2 | XP_015626114.1 | 2 - 3 | 546 | 66 | ||
| GenBank mRNA | AK065045.1 | - | 2 - 3 | 548 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770627.1, XM_015770628.2 transcripts.
Set Membership: |
|

LOC4330499
Genomic Map