Assay Details
| Gene Symbol: LOC4330499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770624.1 | XP_015626110.1 | 5 - 6 | 777 | 65 | |
| XM_015770625.1 | XP_015626111.1 | 5 - 6 | 776 | 65 | ||
| XM_015770627.1 | XP_015626113.1 | 4 - 5 | 265 | 65 | ||
| XM_015770628.2 | XP_015626114.1 | 5 - 6 | 740 | 65 | ||
| GenBank mRNA | AK065045.1 | - | 5 - 6 | 742 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770627.1, XM_015770628.2 transcripts.
Set Membership: |
|

LOC4330499
Genomic Map