Assay Details
Gene Symbol: LOC4330560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767435.1 | XP_015622921.1 | 1 - 2 | 447 | 125 | |
GenBank mRNA | AK061514.1 | - | 1 - 2 | 359 | 125 | |
AK104061.1 | - | 1 - 2 | 297 | 125 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015767435.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
