Assay Details
| Gene Symbol: LOC4330597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771384.2 | XP_015626870.1 | 1 - 2 | 201 | 69 | |
| GenBank mRNA | AK121427.1 | - | 1 - 2 | 143 | 69 | |
| BT130634.1 | - | 1 - 2 | 128 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771384.2 transcripts.
Set Membership: |
|

LOC4330597
Genomic Map