Assay Details
| Gene Symbol: LOC4330597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771384.2 | XP_015626870.1 | 2 - 3 | 357 | 81 | |
| GenBank mRNA | AK121427.1 | - | 2 - 3 | 299 | 81 | |
| BT130634.1 | - | 2 - 3 | 284 | 81 | ||
| EU605823.1 | - | 1 - 2 | 149 | 81 | ||
More Information
Set Membership: |
|

LOC4330597
Genomic Map