Assay Details
| Gene Symbol: LOC4326075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787702.2 | XP_015643188.1 | 18 - 19 | 3537 | 118 | |
| GenBank mRNA | AK242944.1 | - | 12 - 13 | 1985 | 118 | |
More Information
Set Membership: |
|

LOC4326075
Genomic Map