Assay Details
| Gene Symbol: LOC4327922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762193.1 | XP_015617679.1 | 9 - 10 | 1479 | 75 | |
| GenBank mRNA | AK067665.1 | - | 9 - 10 | 1379 | 75 | |
More Information
Set Membership: |
|

LOC4327922
Genomic Map